missing translation for 'onlineSavingsMsg'
Learn More

Tributylmethylammonium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

Catalog No. T26795G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26795G 5 g
T267925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26795G Supplier TCI America Supplier No. T26795G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 405514-94-5
Molecular Formula C15H30F6N2O4S2
Molecular Weight (g/mol) 480.53
MDL Number MFCD12761447
InChI Key XALVHDZWUBSWES-UHFFFAOYSA-N
PubChem CID 44630019
IUPAC Name tributyl(methyl)azanium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
SMILES CCCC[N+](C)(CCCC)CCCC.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 480.53
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Tributylmethylammonium Bis(trifluoromethanesulfonyl)imide