missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributylamine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | -70°C |
| Color | Yellow |
| Boiling Point | 217°C |
| Quantity | 500 mL |
| UN Number | 2542 |
| Formula Weight | 185.36 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | Tributylamine |
Safety and Handling
| EINECSNumber | (2)-0142 |
| RTECSNumber | YA0350000 |
| TSCA | Yes |