accessibility menu, dialog, popup

UserName

CAS RN 102-82-9

IUPAC Name: tributylamine
Synonyms: tributylamine tributilamina tri N-butylamine tris[N-butylamine]
Molecular Weight (g/mol): 185.36
Molecular Formula: C12H27N
InChi Key: IMFACGCPASFAPR-UHFFFAOYSA-N
SMILES: CCCCN(CCCC)CCCC
Boiling Point 
  • (4)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (7)
  • (4)
  • (3)
  • (2)
  • (4)
  • (5)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)