missing translation for 'onlineSavingsMsg'
Learn More

Tributylamine 98.0+%, TCI America™

Catalog No. t035725ml
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T035725ML 25 mL
T0357500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T035725ML Supplier TCI America Supplier No. T035725ML
Only null left

Chemical Identifiers

CAS 102-82-9
Molecular Formula C12H27N
Molecular Weight (g/mol) 185.355
MDL Number MFCD00009431
InChI Key IMFACGCPASFAPR-UHFFFAOYSA-N
Synonym tributylamine, tri-n-butylamine, 1-butanamine, n,n-dibutyl, n,n-dibutyl-1-butanamine, tributilamina, tris-n-butylamine, amine, tributyl, tris n-butylamine, tributilamina romanian, unii-c3tzb2w0r7
PubChem CID 7622
ChEBI CHEBI:38905
IUPAC Name N,N-dibutylbutan-1-amine
SMILES CCCCN(CCCC)CCCC

Specifications

Melting Point -70°C
Color Yellow
Boiling Point 217°C
Quantity 25 mL
UN Number 2542
Formula Weight 185.36
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Tributylamine
EINECSNumber (2)-0142
RTECSNumber YA0350000
TSCA Yes