Learn More
Tributyl phosphate, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 169930025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Tributyl phosphate | |
| 126-73-8 | |
| 0.9790g/mL | |
| 146°C | |
| 99% min. (GC) | |
| C12H27O4P | |
| [CH3(CH2)3O]3P(O) | |
| 2.5 L | |
| 0.979 | |
| tri-n-butyl phosphate, tributylphosphate, butyl phosphate, phosphoric acid tributyl ester, tributylphosphat, celluphos 4, disflamoll tb, tributilfosfato, tributylfosfaat, tributyle phosphate de | |
| STCOOQWBFONSKY-UHFFFAOYSA-N | |
| tributyl phosphate | |
| 31357 | |
| CHEBI:35019 | |
| 99+% |
| 99% | |
| -79.0°C | |
| 289.0°C | |
| Authentic | |
| Glass Bottle | |
| 1.4230 to 1.4250 | |
| MFCD00009436 | |
| 01,II,397 | |
| 15,9781 | |
| Solubility in water: 0.6g/100mL (20°C). Other solubilities: soluble in alcohols,ketones,ethers,benzene,,carbon disulfide and most organic solvents | |
| CCCCOP(=O)(OCCCC)OCCCC | |
| 266.32 | |
| 3.5 mPa.s (25°C) | |
| 266.32 | |
| Liquid |
Chemical Identifiers
| 126-73-8 | |
| 266.32 | |
| STCOOQWBFONSKY-UHFFFAOYSA-N | |
| 31357 | |
| tributyl phosphate |
| C12H27O4P | |
| MFCD00009436 | |
| tri-n-butyl phosphate, tributylphosphate, butyl phosphate, phosphoric acid tributyl ester, tributylphosphat, celluphos 4, disflamoll tb, tributilfosfato, tributylfosfaat, tributyle phosphate de | |
| CHEBI:35019 | |
| CCCCOP(=O)(OCCCC)OCCCC |
Safety and Handling
GHS H Statement
Causes skin irritation.
Suspected of causing cancer.
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Obtain special instructions before use.
Wear pro
GHS Signal Word: Warning
EINECSNumber : 204-800-2
RTECSNumber : TC7700000
TSCA : TSCA
RUO – Research Use Only