missing translation for 'onlineSavingsMsg'
Learn More

Tributyl phosphate, 99+%, Thermo Scientific Chemicals

Catalog No. AC169930100
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Metal drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169930100 10 L Metal drum
AC169930010 1 L Glass Bottle
AC169930025 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC169930100 Supplier Thermo Scientific Chemicals Supplier No. 169930100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 126-73-8
Molecular Formula C12H27O4P
Molecular Weight (g/mol) 266.32
MDL Number MFCD00009436
InChI Key STCOOQWBFONSKY-UHFFFAOYSA-N
Synonym tri-n-butyl phosphate, tributylphosphate, butyl phosphate, phosphoric acid tributyl ester, tributylphosphat, celluphos 4, disflamoll tb, tributilfosfato, tributylfosfaat, tributyle phosphate de
PubChem CID 31357
ChEBI CHEBI:35019
IUPAC Name tributyl phosphate
SMILES CCCCOP(=O)(OCCCC)OCCCC

Specifications

Melting Point -79.0°C
Density 0.9790g/mL
Boiling Point 289.0°C
Flash Point 146°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Metal drum
Refractive Index 1.4230 to 1.4250
Linear Formula [CH3(CH2)3O]3P(O)
Quantity 10 L
Beilstein 01,II,397
Merck Index 15,9781
Specific Gravity 0.979
Solubility Information Solubility in water: 0.6g/100mL (20°C). Other solubilities: soluble in alcohols,ketones,ethers,benzene,,carbon disulfide and most organic solvents
Viscosity 3.5 mPa.s (25°C)
Formula Weight 266.32
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Tributyl phosphate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Suspected of causing cancer.
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Obtain special instructions before use.
Wear pro
EINECSNumber 204-800-2
RTECSNumber TC7700000
TSCA TSCA

RUO – Research Use Only