missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributyl Phosphate, 99%+, Reagents
Tributyl Phosphate, 99%+, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C23414001A
Specifications
| Tributyl Phosphate, 99%+ | |
| 289°C | |
| Colorless to Slight Yellow | |
| -80°C | |
| C12H27O4P | |
| 0.98 | |
| STCOOQWBFONSKY-UHFFFAOYSA-N | |
| tributyl phosphate | |
| Reagent |
| 126-73-8 | |
| -80°C | |
| 289°C | |
| Poly Bottle | |
| 1 L | |
| Tri-n-butyl Phosphate, Butyl Phosphate | |
| CCCCOP(=O)(OCCCC)OCCCC | |
| 266.32 | |
| Liquid |
Chemical Identifiers
| 126-73-8 | |
| 266.32 | |
| Tri-n-butyl Phosphate, Butyl Phosphate | |
| CCCCOP(=O)(OCCCC)OCCCC |
| C12H27O4P | |
| STCOOQWBFONSKY-UHFFFAOYSA-N | |
| tributyl phosphate |