missing translation for 'onlineSavingsMsg'
Learn More

Tributyl Phosphate, 98.5%+, Reagents

Catalog No. 01336915
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
15 gal.
200 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336915 20 L
01336918 15 gal.
01336917 1 L
01336916 200 L
01336914 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01336915 Supplier Labchem Inc Supplier No. C234150020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tributyl Phosphate, 98.5%+, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 126-73-8
Molecular Formula C12H27O4P
Molecular Weight (g/mol) 266.32
InChI Key STCOOQWBFONSKY-UHFFFAOYSA-N
Synonym Tri-n-butyl Phosphate, Butyl Phosphate
IUPAC Name tributyl phosphate
SMILES CCCCOP(=O)(OCCCC)OCCCC

Specifications

Boiling Range 289°C
Melting Point -80°C
Color Colorless to Slight Yellow
Boiling Point 289°C
Melting Range -80°C
Packaging Cubitainer
Quantity 20 L
Specific Gravity 0.98
Grade Reagent
Physical Form Liquid
Chemical Name or Material Tributyl Phosphate, 98.5%+
Show More Show Less