missing translation for 'onlineSavingsMsg'
Learn More

Tributyl(ethyl)phosphonium Diethyl Phosphate 96.0+%, TCI America™

Catalog No. T36415G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T36415G 5 g
T364125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T36415G Supplier TCI America Supplier No. T36415G
Only null left

Chemical Identifiers

CAS 20445-94-7
Molecular Formula C18H42O4P2
Molecular Weight (g/mol) 384.478
MDL Number MFCD17018975
InChI Key RDCTYOLAEUHRGG-UHFFFAOYSA-M
PubChem CID 60196373
IUPAC Name diethyl phosphate;tributyl(ethyl)phosphanium
SMILES CCCC[P+](CC)(CCCC)CCCC.CCOP(=O)([O-])OCC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 384.48
Percent Purity ≥96.0% (T)
Physical Form Liquid
Chemical Name or Material Tributyl(ethyl)phosphonium Diethyl Phosphate