missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributyl Citrate 98.0+%, TCI America™
$32.72 - $86.64
Chemical Identifiers
| CAS | 77-94-1 |
|---|---|
| Molecular Formula | C18H32O7 |
| Molecular Weight (g/mol) | 360.45 |
| MDL Number | MFCD00027217 |
| InChI Key | ZFOZVQLOBQUTQQ-UHFFFAOYSA-N |
| Synonym | tributyl citrate, butyl citrate, tri-n-butyl citrate, n-butyl citrate, citroflex 4, citric acid, tributyl ester, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, tributyl ester, butyl citrate van, unii-827d5b1b6s, 2-hydroxy-1,2,3-propanetricarboxylic acid, tributyl ester |
| PubChem CID | 6507 |
| IUPAC Name | 1,2,3-tributyl 2-hydroxypropane-1,2,3-tricarboxylate |
| SMILES | CCCCOC(=O)CC(O)(CC(=O)OCCCC)C(=O)OCCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C036625ML
|
TCI America
C036625ML |
25 mL |
Each for $32.72
|
|
|||||
|
C0366500ML
|
TCI America
C0366500ML |
500 mL |
Each for $86.64
|
|
|||||
Chemical Identifiers
| 77-94-1 | |
| 360.45 | |
| ZFOZVQLOBQUTQQ-UHFFFAOYSA-N | |
| 6507 | |
| CCCCOC(=O)CC(O)(CC(=O)OCCCC)C(=O)OCCCC |
| C18H32O7 | |
| MFCD00027217 | |
| tributyl citrate, butyl citrate, tri-n-butyl citrate, n-butyl citrate, citroflex 4, citric acid, tributyl ester, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, tributyl ester, butyl citrate van, unii-827d5b1b6s, 2-hydroxy-1,2,3-propanetricarboxylic acid, tributyl ester | |
| 1,2,3-tributyl 2-hydroxypropane-1,2,3-tricarboxylate |
Specifications
| 77-94-1 | |
| Yellow | |
| C18H32O7 | |
| 25 mL | |
| ZFOZVQLOBQUTQQ-UHFFFAOYSA-N | |
| 1,2,3-tributyl 2-hydroxypropane-1,2,3-tricarboxylate | |
| 6507 | |
| ≥98.0% (GC) | |
| Tributyl Citrate |
| -20°C | |
| 233°C | |
| MFCD00027217 | |
| tributyl citrate, butyl citrate, tri-n-butyl citrate, n-butyl citrate, citroflex 4, citric acid, tributyl ester, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, tributyl ester, butyl citrate van, unii-827d5b1b6s, 2-hydroxy-1,2,3-propanetricarboxylic acid, tributyl ester | |
| CCCCOC(=O)CC(O)(CC(=O)OCCCC)C(=O)OCCCC | |
| 360.45 | |
| 360.45 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-1320
RTECSNumber : TZ8608000
TSCA : Yes