missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributyl Citrate 98.0+%, TCI America™
Supplier: TCI America C036625ML
Specifications
| Tributyl Citrate | |
| -20°C | |
| 233°C | |
| MFCD00027217 | |
| tributyl citrate, butyl citrate, tri-n-butyl citrate, n-butyl citrate, citroflex 4, citric acid, tributyl ester, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, tributyl ester, butyl citrate van, unii-827d5b1b6s, 2-hydroxy-1,2,3-propanetricarboxylic acid, tributyl ester | |
| CCCCOC(=O)CC(O)(CC(=O)OCCCC)C(=O)OCCCC | |
| 360.45 | |
| 360.45 | |
| Liquid |
| 77-94-1 | |
| Yellow | |
| C18H32O7 | |
| 25 mL | |
| ZFOZVQLOBQUTQQ-UHFFFAOYSA-N | |
| 1,2,3-tributyl 2-hydroxypropane-1,2,3-tricarboxylate | |
| 6507 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 77-94-1 | |
| 360.45 | |
| ZFOZVQLOBQUTQQ-UHFFFAOYSA-N | |
| 6507 | |
| CCCCOC(=O)CC(O)(CC(=O)OCCCC)C(=O)OCCCC |
| C18H32O7 | |
| MFCD00027217 | |
| tributyl citrate, butyl citrate, tri-n-butyl citrate, n-butyl citrate, citroflex 4, citric acid, tributyl ester, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, tributyl ester, butyl citrate van, unii-827d5b1b6s, 2-hydroxy-1,2,3-propanetricarboxylic acid, tributyl ester | |
| 1,2,3-tributyl 2-hydroxypropane-1,2,3-tricarboxylate |
Safety and Handling
EINECSNumber : (2)-1320
RTECSNumber : TZ8608000
TSCA : Yes