missing translation for 'onlineSavingsMsg'
Learn More

Tributyl citrate, 99+%

CAS: 77-94-1 | C18H32O7 | 360.45 g/mol

Supplier:  Thermo Scientific Chemicals 421440010

 View more versions of this product

Catalog No. AC421440010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tributyl citrate
0.02% max. (as citric acid)
1.0420g/mL
157°C
99% min. (GC)
C18H32O7
HOCCOO(CH2)3CH3(CH2COO(CH2)3CH3)2
1 kg
15, 1565
Solubility in water: insoluble. Other solubilities: miscible with most organic liquids,soluble in ethanol and ether
CCCCOC(=O)CC(O)(CC(=O)OCCCC)C(=O)OCCCC
360.45
31.9 mPa.s (25°C)
99+%
77-94-1
-20°C
325°C
Authentic
Glass bottle
1.4430 to 1.446
MFCD00027217
1.042
tributyl citrate, butyl citrate, tri-n-butyl citrate, n-butyl citrate, citroflex 4, citric acid, tributyl ester, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, tributyl ester, butyl citrate van, unii-827d5b1b6s, 2-hydroxy-1,2,3-propanetricarboxylic acid, tributyl ester
ZFOZVQLOBQUTQQ-UHFFFAOYSA-N
tributyl 2-hydroxypropane-1,2,3-tricarboxylate
6507
360.45
Liquid

Chemical Identifiers

77-94-1
360.45
ZFOZVQLOBQUTQQ-UHFFFAOYSA-N
6507
CCCCOC(=O)CC(O)(CC(=O)OCCCC)C(=O)OCCCC
C18H32O7
MFCD00027217
tributyl citrate, butyl citrate, tri-n-butyl citrate, n-butyl citrate, citroflex 4, citric acid, tributyl ester, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, tributyl ester, butyl citrate van, unii-827d5b1b6s, 2-hydroxy-1,2,3-propanetricarboxylic acid, tributyl ester
tributyl 2-hydroxypropane-1,2,3-tricarboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 201-071-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only