missing translation for 'onlineSavingsMsg'
Learn More

Tributyl citrate, 99+%

Catalog No. p-3737611
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC421442500 250 g Glass bottle
AC421440010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC421442500 Supplier Thermo Scientific Chemicals Supplier No. 421442500

CAS: 77-94-1 | C18H32O7 | 360.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 77-94-1
Molecular Formula C18H32O7
Molecular Weight (g/mol) 360.45
MDL Number MFCD00027217
InChI Key ZFOZVQLOBQUTQQ-UHFFFAOYSA-N
Synonym tributyl citrate, butyl citrate, tri-n-butyl citrate, n-butyl citrate, citroflex 4, citric acid, tributyl ester, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, tributyl ester, butyl citrate van, unii-827d5b1b6s, 2-hydroxy-1,2,3-propanetricarboxylic acid, tributyl ester
PubChem CID 6507
IUPAC Name tributyl 2-hydroxypropane-1,2,3-tricarboxylate
SMILES CCCCOC(=O)CC(O)(CC(=O)OCCCC)C(=O)OCCCC

Specifications

Acidity 0.02% max. (as citric acid)
Melting Point -20°C
Density 1.0420g/mL
Boiling Point 325°C
Flash Point 157°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4430 to 1.446
Linear Formula HOCCOO(CH2)3CH3(CH2COO(CH2)3CH3)2
Quantity 250 g
Specific Gravity 1.042
Merck Index 15, 1565
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with most organic liquids,soluble in ethanol and ether
Viscosity 31.9 mPa.s (25°C)
Formula Weight 360.45
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Tributyl citrate
Show More Show Less
EINECSNumber 201-071-2

RUO – Research Use Only