missing translation for 'onlineSavingsMsg'
Learn More

Tributyl(1,3-dioxolan-2-ylmethyl)phosphonium Bromide 98.0+%, TCI America™

Catalog No. T27185G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T27185G Supplier TCI America Supplier No. T27185G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 115754-62-6
Molecular Formula C16H34BrO2P
Molecular Weight (g/mol) 369.32
MDL Number MFCD09859807
InChI Key SABIECHYMHKXJI-UHFFFAOYSA-M
PubChem CID 22292430
IUPAC Name tributyl[(1,3-dioxolan-2-yl)methyl]phosphanium bromide
SMILES [Br-].CCCC[P+](CCCC)(CCCC)CC1OCCO1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 369.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material Tributyl(1,3-dioxolan-2-ylmethyl)phosphonium Bromide
TSCA No
Recommended Storage Refrigerator