accessibility menu, dialog, popup

UserName

H3CPCH3CH3OOBr

CAS RN 115754-62-6

IUPAC Name: tributyl[(1,3-dioxolan-2-yl)methyl]phosphanium bromide
Synonyms: tributyl(1,3-dioxolan-2-ylmethyl)phosphanium bromide
Molecular Weight (g/mol): 369.32
Molecular Formula: C16H34BrO2P
InChi Key: SABIECHYMHKXJI-UHFFFAOYSA-M
SMILES: [Br-].CCCC[P+](CCCC)(CCCC)CC1OCCO1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results