missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tribromoethylene 97.0+%, TCI America™
$124.89 - $605.83
Chemical Identifiers
| CAS | 598-16-3 |
|---|---|
| Molecular Formula | C2HBr3 |
| Molecular Weight (g/mol) | 264.74 |
| MDL Number | MFCD00039164 |
| InChI Key | OZVJKTHTULCNHB-UHFFFAOYSA-N |
| PubChem CID | 11712 |
| IUPAC Name | 1,1,2-tribromoethene |
| SMILES | BrC=C(Br)Br |
Chemical Identifiers
| 598-16-3 | |
| 264.74 | |
| OZVJKTHTULCNHB-UHFFFAOYSA-N | |
| 1,1,2-tribromoethene |
| C2HBr3 | |
| MFCD00039164 | |
| 11712 | |
| BrC=C(Br)Br |
Specifications
| 598-16-3 | |
| 163°C | |
| MFCD00039164 | |
| OZVJKTHTULCNHB-UHFFFAOYSA-N | |
| 1,1,2-tribromoethene | |
| 11712 | |
| ≥97.0% (GC) | |
| Tribromoethylene |
| Yellow | |
| C2HBr3 | |
| 25 g | |
| BrC=C(Br)Br | |
| 264.74 | |
| 264.74 | |
| Liquid |
Safety and Handling
RTECSNumber : KX4300000
TSCA : Yes