missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tribenzylamine 99.0+%, TCI America™
$41.02 - $272.16
Chemical Identifiers
| CAS | 620-40-6 |
|---|---|
| Molecular Formula | C21H21N |
| Molecular Weight (g/mol) | 287.41 |
| MDL Number | MFCD00004773 |
| InChI Key | MXHTZQSKTCCMFG-UHFFFAOYSA-N |
| Synonym | tribenzylamine, benzenemethanamine, n,n-bis phenylmethyl, unii-hz10o1931j, trisbenzylamine, n,n-dibenzyl-1-phenyl-methanamine, pubchem21076, acmc-1atok, dsstox_cid_27031, dsstox_rid_82052 |
| PubChem CID | 24321 |
| IUPAC Name | tribenzylamine |
| SMILES | C(N(CC1=CC=CC=C1)CC1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 620-40-6 | |
| 287.41 | |
| MXHTZQSKTCCMFG-UHFFFAOYSA-N | |
| 24321 | |
| C(N(CC1=CC=CC=C1)CC1=CC=CC=C1)C1=CC=CC=C1 |
| C21H21N | |
| MFCD00004773 | |
| tribenzylamine, benzenemethanamine, n,n-bis phenylmethyl, unii-hz10o1931j, trisbenzylamine, n,n-dibenzyl-1-phenyl-methanamine, pubchem21076, acmc-1atok, dsstox_cid_27031, dsstox_rid_82052 | |
| tribenzylamine |
Specifications
| 620-40-6 | |
| White | |
| C21H21N | |
| 25 g | |
| MXHTZQSKTCCMFG-UHFFFAOYSA-N | |
| tribenzylamine | |
| 24321 | |
| ≥99.0% (GC,T) | |
| Tribenzylamine |
| 93°C | |
| 390°C | |
| MFCD00004773 | |
| tribenzylamine, benzenemethanamine, n,n-bis phenylmethyl, unii-hz10o1931j, trisbenzylamine, n,n-dibenzyl-1-phenyl-methanamine, pubchem21076, acmc-1atok, dsstox_cid_27031, dsstox_rid_82052 | |
| C(N(CC1=CC=CC=C1)CC1=CC=CC=C1)C1=CC=CC=C1 | |
| 287.41 | |
| 287.41 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-0353
TSCA : Yes