missing translation for 'onlineSavingsMsg'
Learn More

Tribenzylamine 99.0+%, TCI America™

Catalog No. T034125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T034125G 25 g
T0341500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T034125G Supplier TCI America Supplier No. T034125G

Chemical Identifiers

CAS 620-40-6
Molecular Formula C21H21N
Molecular Weight (g/mol) 287.41
MDL Number MFCD00004773
InChI Key MXHTZQSKTCCMFG-UHFFFAOYSA-N
Synonym tribenzylamine, benzenemethanamine, n,n-bis phenylmethyl, unii-hz10o1931j, trisbenzylamine, n,n-dibenzyl-1-phenyl-methanamine, pubchem21076, acmc-1atok, dsstox_cid_27031, dsstox_rid_82052
PubChem CID 24321
IUPAC Name tribenzylamine
SMILES C(N(CC1=CC=CC=C1)CC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 93°C
Color White
Boiling Point 390°C
Quantity 25 g
Formula Weight 287.41
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tribenzylamine
EINECSNumber (3)-0353
TSCA Yes