missing translation for 'onlineSavingsMsg'
Learn More

Tribehenin 90.0+%, TCI America™

$143.54 - $423.14

Chemical Identifiers

CAS 18641-57-1
Molecular Formula C69H134O6
Molecular Weight (g/mol) 1059.825
MDL Number MFCD00056274
InChI Key DMBUODUULYCPAK-UHFFFAOYSA-N
Synonym Glycerol Tridocosanoate, 1,2,3-Tridocosanoyl Glycerol
PubChem CID 62726
IUPAC Name 2,3-di(docosanoyloxy)propyl docosanoate
SMILES CCCCCCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCCCCCCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T13911G
View Documents
TCI America
T13911G
1 g N/A
Only null left
 
T13915G
View Documents
TCI America
T13915G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

18641-57-1
1059.825
DMBUODUULYCPAK-UHFFFAOYSA-N
62726
CCCCCCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCCCCCCCC
C69H134O6
MFCD00056274
Glycerol Tridocosanoate, 1,2,3-Tridocosanoyl Glycerol
2,3-di(docosanoyloxy)propyl docosanoate

Specifications

18641-57-1
White
MFCD00056274
Glycerol Tridocosanoate, 1,2,3-Tridocosanoyl Glycerol
CCCCCCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCCCCCCCC
1059.825
1059.83
Crystalline Powder
83°C
C69H134O6
1 g
DMBUODUULYCPAK-UHFFFAOYSA-N
2,3-di(docosanoyloxy)propyl docosanoate
62726
≥90.0% (T)
Tribehenin
Videos
Safety and Handling

Safety and Handling

RTECSNumber : JR1305000

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'