accessibility menu, dialog, popup

UserName

CAS RN 18641-57-1

H3COOOOCH3OOCH3

CAS RN 18641-57-1

IUPAC Name: 1,3-bis(docosanoyloxy)propan-2-yl docosanoate
Synonyms: tribehenin tridocosanoin docosanoin, tri- (7CI,8CI) tribehenoyl glycerol glyceryl tribehenate compritol 888 tri- (7CI,8CI) docosanoin
Molecular Weight (g/mol): 1059.83
Molecular Formula: C69H134O6
InChi Key: DMBUODUULYCPAK-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCCCCCCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results