missing translation for 'onlineSavingsMsg'
Learn More

Tribehenin 90.0+%, TCI America™

Catalog No. T13911G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13911G 1 g
T13915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T13911G Supplier TCI America Supplier No. T13911G
Only null left

Chemical Identifiers

CAS 18641-57-1
Molecular Formula C69H134O6
Molecular Weight (g/mol) 1059.825
MDL Number MFCD00056274
InChI Key DMBUODUULYCPAK-UHFFFAOYSA-N
Synonym Glycerol Tridocosanoate, 1,2,3-Tridocosanoyl Glycerol
PubChem CID 62726
IUPAC Name 2,3-di(docosanoyloxy)propyl docosanoate
SMILES CCCCCCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 83°C
Color White
Quantity 1 g
Formula Weight 1059.83
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tribehenin
RTECSNumber JR1305000
TSCA No