missing translation for 'onlineSavingsMsg'
Learn More

Triamterene Related Compound C Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111002268
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002268 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002268 Supplier Merck Emd Millipore Supplier No. PHR1820100MG
Only null left
Triamterene Related Compound C is a pharmaceutical impurity of a potassium-sparing diuretic, triamterene. Triamterene exhibits antihypertensive activity. It is used in combination with thiazide deuretic, such as benzthiazide for treating hypertension and edema associated with mild to moderate congestive heart failure.

Specifications

CAS 19152-93-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C12H10N6 O
Synonym 2,4-Diamino-7-hydroxy-6-phenylpteridine; 2,4-diamino-7-hydroxy-6-phenylpteridine; NSC 33416
Type Certified Reference Material
Quantity 100 mg
Formula Weight 254.25
Grade Certified Reference Material