missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triamterene Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 396-01-0 |
| Linear Formula | C12H11N7 |
| MDL Number | MFCD00006708 |
| Synonym | Triamterene; 2,4,7-Triamino-6-phenylpteridine; 6-Phenyl-2,4,7-pteridinetriamine |
| Type | Certified Reference Material |
| Quantity | 400 mg |
| Formula Weight | 253.26 |
| Grade | Certified Reference Material |