missing translation for 'onlineSavingsMsg'
Learn More

Triamcinolone acetonide, Analytical Standard, ≥99%, MilliporeSigma™ Supelco™

Catalog No. p-7193503
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015455 250 mg
111015453 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015455 Supplier Merck Emd Millipore Supplier No. T6501250MG
Only null left

Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. T6501.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.

Specifications

Packaging Glass Bottle
CAS 76-25-5
Physical Form Neat
Linear Formula C24H31FO6
MDL Number MFCD00056834
Synonym 9 alpha-Fluoro-11 beta,16 alpha,17 alpha,21-tetrahydroxy-1,4-pregnadiene-3,20-dione 16,17-acetonide; 9 alpha-Fluoro-16 alpha-hydroxyprednisolone 16 alpha,17 alpha-acetonide
Quantity 250 mg
Formula Weight 434.50
Percent Purity ≥99%
Grade Analytical Standard