Learn More
Triamcinolone acetonide, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J6354803
| Quantity | 1 g |
|---|
Description
Triamcinolone acetonide suppresses allergic, inflammatory, and immune responses. Intravitreal administration of triamcinolone acetonide is used against diabetic macular edema. It is also used to relieve the discomfort of mouth sores.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in DMSO or ethanol. Slightly soluble in water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, acids, heat.
Chemical Identifiers
| 76-25-5 | |
| 434.504 | |
| YNDXUCZADRHECN-JNQJZLCISA-N | |
| 6436 | |
| CC1(OC2CC3C4CCC5=CC(=O)C=CC5(C4(C(CC3(C2(O1)C(=O)CO)C)O)F)C)C |
| C24H31FO6 | |
| MFCD00056834 | |
| triamcinolone acetonide, azmacort, nasacort aq, aristoderm, aristogel, kenalone, solodelf, tramacin, triaceton, tricinolon | |
| CHEBI:71418 |
Specifications
| Triamcinolone acetonide | |
| 274°C to 278°C | |
| C24H31FO6 | |
| MFCD00056834 | |
| triamcinolone acetonide, azmacort, nasacort aq, aristoderm, aristogel, kenalone, solodelf, tramacin, triaceton, tricinolon | |
| YNDXUCZADRHECN-JNQJZLCISA-N | |
| 434.504 | |
| CHEBI:71418 | |
| ≥98% |
| 76-25-5 | |
| White | |
| 1 g | |
| 14,9596 | |
| Soluble in DMSO or ethanol; Slightly soluble in water. | |
| CC1(OC2CC3C4CCC5=CC(=O)C=CC5(C4(C(CC3(C2(O1)C(=O)CO)C)O)F)C)C | |
| 6436 | |
| 434.5 | |
| Powder |
Safety and Handling
GHS H Statement
H360-H302
May damage fertility or the unborn child.
Harmful if swallowed.
P201-P202-P264b-P270-P280i-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
H302-H315-H319-H360
EINECSNumber : 200-948-7
RTECSNumber : TU3920000
TSCA : Yes
Recommended Storage : Keep cold