accessibility menu, dialog, popup

UserName

CAS RN 76-25-5

IUPAC Name: (1S,2S,4R,8S,9S,11S,12R,13S)-12-fluoro-11-hydroxy-8-(2-hydroxyacetyl)-6,6,9,13-tetramethyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-16-one
Synonyms: nasacort AQ triamcinolone acetonide acetospan trivaris tricinolon omcilon A tricort40 A, kenacort triamcincolone acetonide triesence
Molecular Weight (g/mol): 434.5
Molecular Formula: C24H31FO6
InChi Key: YNDXUCZADRHECN-JNQJZLCISA-N
SMILES: CC1(C)O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO
Percent Purity 
  • (2)
  • (3)
Grade 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)