missing translation for 'onlineSavingsMsg'
Learn More

Triallylamine 95.0+%, TCI America™

Catalog No. p-7136224
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T033225ML 25 mL
T0332500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T033225ML Supplier TCI America Supplier No. T033225ML
Only null left

Chemical Identifiers

CAS 102-70-5
Molecular Formula C9H15N
Molecular Weight (g/mol) 137.226
MDL Number MFCD00026093
InChI Key VPYJNCGUESNPMV-UHFFFAOYSA-N
Synonym triallylamine, tris 2-propenyl amine, 2-propen-1-amine, n,n-di-2-propenyl, triallyl amine, unii-b6n19xc04r, ccris 4876, n,n-diallylprop-2-en-1-amine, n,n-di-2-propenyl-2-propen-1-amine, ch2=chch2 3n, 4-04-00-01061 beilstein handbook reference
PubChem CID 7617
IUPAC Name N,N-bis(prop-2-enyl)prop-2-en-1-amine
SMILES C=CCN(CC=C)CC=C

Specifications

Boiling Point 156°C
Quantity 25 mL
UN Number 2610
Formula Weight 137.23
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Triallylamine
EINECSNumber (2)-2381
RTECSNumber XX5950000
TSCA Yes