accessibility menu, dialog, popup

UserName

CH2NH2CCH2

CAS RN 102-70-5

IUPAC Name: tris(prop-2-en-1-yl)amine
Synonyms: triallylamine
Molecular Weight (g/mol): 137.23
Molecular Formula: C9H15N
InChi Key: VPYJNCGUESNPMV-UHFFFAOYSA-N
SMILES: C=CCN(CC=C)CC=C
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)