missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triallylamine 95.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 156°C |
| Quantity | 500 mL |
| UN Number | 2610 |
| Formula Weight | 137.23 |
| Percent Purity | ≥95.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Liquid |
| Chemical Name or Material | Triallylamine |
Safety and Handling
| EINECSNumber | (2)-2381 |
| RTECSNumber | XX5950000 |
| TSCA | Yes |