missing translation for 'onlineSavingsMsg'
Learn More

Triallyl-s-triazine-2,4,6(1H,3H,5H)-trione, 98%, Stabilized

Catalog No. p-202352
Change view
Click to view available options
Quantity:
100 g
Catalog No. Quantity
AC342121000 100 g
1 options

Catalog No. AC342121000

Supplier: Thermo Scientific Chemicals 342121000

Only null left

CAS: 1025-15-6 | C12H15N3O3 | 249.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1025-15-6
Molecular Formula C12H15N3O3
Molecular Weight (g/mol) 249.27
MDL Number MFCD00006554
InChI Key KOMNUTZXSVSERR-UHFFFAOYSA-N
Synonym triallyl isocyanurate, isocyanuric acid triallyl ester, 1,3,5-triallyl-1,3,5-triazinane-2,4,6-trione, triallylisocyanurate, taic, diak 7, 1,3,5-triallylisocyanurate, 1,3,5-triallyl isocyanurate, 1,3,5-triallylisocyanuric acid, 1,3,5-tri-2-propenyl-1,3,5-triazine-2,4,6 1h,3h,5h-trione
PubChem CID 13931
IUPAC Name 1,3,5-tris(prop-2-enyl)-1,3,5-triazinane-2,4,6-trione
SMILES C=CCN1C(=O)N(C(=O)N(C1=O)CC=C)CC=C

Specifications

Melting Point 24.8°C
Color Colorless to Yellow
Density 1.1590g/mL
Boiling Point 149.0°C to 152.0°C (4.0 mmHg)
Flash Point 245°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5120 to 1.5140
Quantity 100 g
Beilstein 26, IV, 637
Specific Gravity 1.159
Solubility Information Solubility in water: >1g/L (20°C). Other solubilities: soluble in acetone,dimethyl sulfoxide,ethanol
Stabilizer 1200ppm MEHQ
Viscosity 90.1 mPa.s (30°C)
Formula Weight 249.27
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material Triallyl-s-triazine-2,4,6(1H,3H,5H)-trione, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
EINECSNumber 213-834-7
RTECSNumber XZ1915000
TSCA TSCA

RUO – Research Use Only