accessibility menu, dialog, popup

UserName

CAS RN 1025-15-6

IUPAC Name: tris(prop-2-en-1-yl)-1,3,5-triazinane-2,4,6-trione
Synonyms: tris(prop-2-en-1-yl)-1,3,5-triazinane-2,4,6-trione
Molecular Weight (g/mol): 249.27
Molecular Formula: C12H15N3O3
InChi Key: KOMNUTZXSVSERR-UHFFFAOYSA-N
SMILES: C=CCN1C(=O)N(CC=C)C(=O)N(CC=C)C1=O
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)