missing translation for 'onlineSavingsMsg'
Learn More

Triallyl Phosphate 96.0+%, TCI America™

Catalog No. P026425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P026425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P026425ML Supplier TCI America Supplier No. P026425ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1623-19-4
Molecular Formula C9H15O4P
Molecular Weight (g/mol) 218.19
MDL Number MFCD00014946
InChI Key XHGIFBQQEGRTPB-UHFFFAOYSA-N
Synonym triallyl phosphate, allyl phosphate, triallyl orthophosphate, phosphoric acid, tri-2-propenyl ester, triallylfosfat, triallylphosphate, phosphoric acid triallyl ester, phosphoric acid, triallyl ester, allyl phosphate c3h5o 3po, triallylfosfat czech
PubChem CID 15390
IUPAC Name tris(prop-2-en-1-yl) phosphate
SMILES C=CCOP(=O)(OCC=C)OCC=C

Specifications

Color Yellow
Boiling Point 113°C
Quantity 25 mL
Formula Weight 218.19
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Triallyl Phosphate
EINECSNumber (2)-1960
RTECSNumber TC8575000
TSCA Yes