accessibility menu, dialog, popup

UserName

H2COPOOH2COH2C

CAS RN 1623-19-4

IUPAC Name: tris(prop-2-en-1-yl) phosphate
Synonyms: triallylphosphate
Molecular Weight (g/mol): 218.19
Molecular Formula: C9H15O4P
InChi Key: XHGIFBQQEGRTPB-UHFFFAOYSA-N
SMILES: C=CCOP(=O)(OCC=C)OCC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results