missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triallyl(phenyl)silane 95.0+%, TCI America™
Supplier: TCI America T35501ML
Specifications
| Triallyl(phenyl)silane | |
| Yellow | |
| C15H20Si | |
| 1 mL | |
| C=CC[Si](CC=C)(CC=C)C1=CC=CC=C1 | |
| 228.41 | |
| 228.41 | |
| Liquid |
| 2633-57-0 | |
| 275°C | |
| MFCD00094138 | |
| QFCLQSLLAYLBCU-UHFFFAOYSA-N | |
| phenyltris(prop-2-en-1-yl)silane | |
| 2733229 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 2633-57-0 | |
| 228.41 | |
| QFCLQSLLAYLBCU-UHFFFAOYSA-N | |
| phenyltris(prop-2-en-1-yl)silane |
| C15H20Si | |
| MFCD00094138 | |
| 2733229 | |
| C=CC[Si](CC=C)(CC=C)C1=CC=CC=C1 |
Safety and Handling
TSCA : No