accessibility menu, dialog, popup

UserName

CH2SiH2CCH2

CAS RN 2633-57-0

IUPAC Name: phenyltris(prop-2-en-1-yl)silane
Synonyms: phenyltris(prop-2-en-1-yl)silane
Molecular Weight (g/mol): 228.41
Molecular Formula: C15H20Si
InChi Key: QFCLQSLLAYLBCU-UHFFFAOYSA-N
SMILES: C=CC[Si](CC=C)(CC=C)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results