missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triallyl(methyl)silane 95.0+%, TCI America™
$150.98 - $490.25
Chemical Identifiers
| CAS | 1112-91-0 |
|---|---|
| Molecular Formula | C10H18Si |
| Molecular Weight (g/mol) | 166.339 |
| MDL Number | MFCD00800604 |
| InChI Key | JFCCVNTYPIUJDJ-UHFFFAOYSA-N |
| Synonym | triallylmethylsilane, triallyl methyl silane, methyltris prop-2-en-1-yl silane, methyltriallylsilane, methyl-tris prop-2-enyl silane, silane,methyltri-2-propen-1-yl |
| PubChem CID | 566299 |
| IUPAC Name | methyl-tris(prop-2-enyl)silane |
| SMILES | C[Si](CC=C)(CC=C)CC=C |
Chemical Identifiers
| 1112-91-0 | |
| 166.339 | |
| JFCCVNTYPIUJDJ-UHFFFAOYSA-N | |
| 566299 | |
| C[Si](CC=C)(CC=C)CC=C |
| C10H18Si | |
| MFCD00800604 | |
| triallylmethylsilane, triallyl methyl silane, methyltris prop-2-en-1-yl silane, methyltriallylsilane, methyl-tris prop-2-enyl silane, silane,methyltri-2-propen-1-yl | |
| methyl-tris(prop-2-enyl)silane |
Specifications
| 1112-91-0 | |
| 180°C | |
| MFCD00800604 | |
| 1993 | |
| JFCCVNTYPIUJDJ-UHFFFAOYSA-N | |
| methyl-tris(prop-2-enyl)silane | |
| 566299 | |
| ≥95.0% (GC) | |
| Triallyl(methyl)silane |
| Yellow | |
| C10H18Si | |
| 1 mL | |
| triallylmethylsilane, triallyl methyl silane, methyltris prop-2-en-1-yl silane, methyltriallylsilane, methyl-tris prop-2-enyl silane, silane,methyltri-2-propen-1-yl | |
| C[Si](CC=C)(CC=C)CC=C | |
| 166.339 | |
| 166.34 | |
| Liquid |