accessibility menu, dialog, popup

UserName

H3CSiH2CCH2H2C

CAS RN 1112-91-0

IUPAC Name: methyltris(prop-2-en-1-yl)silane
Synonyms: methyltris(prop-2-en-1-yl)silane
Molecular Weight (g/mol): 166.34
Molecular Formula: C10H18Si
InChi Key: JFCCVNTYPIUJDJ-UHFFFAOYSA-N
SMILES: C[Si](CC=C)(CC=C)CC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results