missing translation for 'onlineSavingsMsg'
Learn More

Triallyl(methyl)silane 95.0+%, TCI America™

Catalog No. T35491ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35491ML 1 mL
T35495ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35491ML Supplier TCI America Supplier No. T35491ML

Chemical Identifiers

CAS 1112-91-0
Molecular Formula C10H18Si
Molecular Weight (g/mol) 166.339
MDL Number MFCD00800604
InChI Key JFCCVNTYPIUJDJ-UHFFFAOYSA-N
Synonym triallylmethylsilane, triallyl methyl silane, methyltris prop-2-en-1-yl silane, methyltriallylsilane, methyl-tris prop-2-enyl silane, silane,methyltri-2-propen-1-yl
PubChem CID 566299
IUPAC Name methyl-tris(prop-2-enyl)silane
SMILES C[Si](CC=C)(CC=C)CC=C

Specifications

Color Yellow
Boiling Point 180°C
Quantity 1 mL
UN Number 1993
Formula Weight 166.34
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Triallyl(methyl)silane