missing translation for 'onlineSavingsMsg'
Learn More

Triallyl Cyanurate 98.0+%, TCI America™

Catalog No. t0333500g
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0333500G 500 g
T033325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0333500G Supplier TCI America Supplier No. T0333500G
Only null left

Chemical Identifiers

CAS 101-37-1
Molecular Formula C12H15N3O3
Molecular Weight (g/mol) 249.27
MDL Number MFCD00006049
InChI Key BJELTSYBAHKXRW-UHFFFAOYSA-N
Synonym triallyl cyanurate, 2,4,6-triallyloxy-1,3,5-triazine, triallyl cyanaurate, activator oc, rhenofit tac, cyanuric acid triallyl ester, perkalink 300, tripropargyl cyanurate, perkalink 300-50d, 1,3,5-triazine, 2,4,6-tris 2-propenyloxy
PubChem CID 7555
IUPAC Name 2,4,6-tris(prop-2-enoxy)-1,3,5-triazine
SMILES C=CCOC1=NC(=NC(=N1)OCC=C)OCC=C

Specifications

Color White
Boiling Point 162°C
Quantity 500 g
UN Number 3077
Formula Weight 249.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Triallyl Cyanurate
EINECSNumber (5)-1059
RTECSNumber XZ2080000
TSCA Yes