accessibility menu, dialog, popup

UserName

H2CONOCH2NOH2CN

CAS RN 101-37-1

IUPAC Name: tris(prop-2-en-1-yloxy)-1,3,5-triazine
Synonyms: tris(prop-2-en-1-yloxy)-1,3,5-triazine
Molecular Weight (g/mol): 249.27
Molecular Formula: C12H15N3O3
InChi Key: BJELTSYBAHKXRW-UHFFFAOYSA-N
SMILES: C=CCOC1=NC(OCC=C)=NC(OCC=C)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results