missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triacontane 98.0+%, TCI America™
$273.38 - $902.90
Chemical Identifiers
| CAS | 638-68-6 |
|---|---|
| Molecular Formula | C30H62 |
| Molecular Weight (g/mol) | 422.826 |
| MDL Number | MFCD00009410 |
| InChI Key | JXTPJDDICSTXJX-UHFFFAOYSA-N |
| Synonym | n-triacontane, triacontane, analytical standard, triacontane, acmc-1b4uf, n-triacontane 250mg, ch3-ch2 28-ch3 |
| PubChem CID | 12535 |
| ChEBI | CHEBI:31006 |
| IUPAC Name | triacontane |
| SMILES | CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC |
Chemical Identifiers
| 638-68-6 | |
| 422.826 | |
| JXTPJDDICSTXJX-UHFFFAOYSA-N | |
| 12535 | |
| triacontane |
| C30H62 | |
| MFCD00009410 | |
| n-triacontane, triacontane, analytical standard, triacontane, acmc-1b4uf, n-triacontane 250mg, ch3-ch2 28-ch3 | |
| CHEBI:31006 | |
| CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC |
Specifications
| 638-68-6 | |
| White | |
| C30H62 | |
| 5 g | |
| JXTPJDDICSTXJX-UHFFFAOYSA-N | |
| triacontane | |
| 12535 | |
| 422.83 | |
| Crystalline Powder |
| 66°C | |
| 200°C | |
| MFCD00009410 | |
| n-triacontane, triacontane, analytical standard, triacontane, acmc-1b4uf, n-triacontane 250mg, ch3-ch2 28-ch3 | |
| CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC | |
| 422.826 | |
| CHEBI:31006 | |
| ≥98.0% (GC) | |
| Triacontane |
Safety and Handling
TSCA : Yes