accessibility menu, dialog, popup

UserName

H3CCH3

CAS RN 638-68-6

IUPAC Name: triacontane
Synonyms: triacontane
Molecular Weight (g/mol): 422.83
Molecular Formula: C30H62
InChi Key: JXTPJDDICSTXJX-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (1)
  • (1)
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (3)
Boiling Point 
  • (1)
  • (4)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (2)