missing translation for 'onlineSavingsMsg'
Learn More

Triacontane, 98%

Catalog No. AC278050010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC278050010 1 g Glass bottle
AC278050050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC278050010 Supplier Thermo Scientific Chemicals Supplier No. 278050010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 638-68-6 | C30H62 | 422.826 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 638-68-6
MDL Number MFCD00009410
InChI Key JXTPJDDICSTXJX-UHFFFAOYSA-N
Synonym n-triacontane, triacontane, analytical standard, triacontane, acmc-1b4uf, n-triacontane 250mg, ch3-ch2 28-ch3
PubChem CID 12535
ChEBI CHEBI:31006
IUPAC Name triacontane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 65°C to 67°C
Color White
Boiling Point 455°C
Flash Point 238°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C30H62
Linear Formula CH3(CH2)28CH3
Quantity 1 g
Beilstein 01, 176
Solubility Information Solubility in water: insoluble
Molecular Weight (g/mol) 422.82
Formula Weight 422.82
Percent Purity 98%
Physical Form Shiny Flakes
Chemical Name or Material Triacontane
Show More Show Less
EINECSNumber 211-349-5

RUO – Research Use Only