missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triacetylresveratrol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 119°C |
| Color | White |
| Quantity | 1 g |
| Synonym | 3,4′C,5-Triacetoxy-trans-stilbene |
| Formula Weight | 354.36 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Triacetylresveratrol |
Safety and Handling
| TSCA | No |