missing translation for 'onlineSavingsMsg'
Learn More

Triacétoxyéthylsilane 96,0+, TCI America™

Numéro de catalogue. E052525G
Modifier l'affichage
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
E052525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. E052525G Fournisseur TCI America Code fournisseur E052525G

Identifiants chimiques

CAS 17689-77-9
Molecular Formula C8H14O6Si
Molecular Weight (g/mol) 234.279
MDL Number MFCD00053703
InChI Key KXJLGCBCRCSXQF-UHFFFAOYSA-N
Synonym ethyltriacetoxysilane, triacetoxyethylsilane, triacetoxy ethyl silane, triacetoxy ethylsilane, ethylsilanetriyl triacetate, silanetriol, ethyl-, triacetate, silanetriol, 1-ethyl-, 1,1,1-triacetate, dsstox_cid_7792, dsstox_rid_78567, dsstox_gsid_27792
PubChem CID 87244
IUPAC Name [acétate de diacétyloxy(éthyl)silyle]
SMILES CC[Si](OC(=O)C)(OC(=O)C)OC(=O)C

Spécifications

Color Jaune
Boiling Point 109°C
Quantity 25 g
UN Number 1760
Formula Weight 234.28
Percent Purity ≥96.0% (GC)
Physical Form Liquide
Chemical Name or Material Triacetoxyethylsilane
EINECSNumber (9)-1939
TSCA Yes