accessibility menu, dialog, popup

UserName

H3CSiOCH3OOH3COOCH3O

CAS RN 17689-77-9

IUPAC Name: bis(acetyloxy)(ethyl)silyl acetate
Synonyms: bis(acetyloxy)(ethyl)silyl acetate
Molecular Weight (g/mol): 234.28
Molecular Formula: C8H14O6Si
InChi Key: KXJLGCBCRCSXQF-UHFFFAOYSA-N
SMILES: CC[Si](OC(C)=O)(OC(C)=O)OC(C)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results