missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triacetin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 102-76-1 |
| Linear Formula | (CH3 COOCH2)2 CHOCOCH3 |
| MDL Number | MFCD00008716 |
| Boiling Point | 258°C to 260°C |
| Synonym | Glyceryl triacetate; 1,2,3-Triacetoxypropane; 1,2,3-Triacetylglycerol; Glycerol triacetate; Triacetin |
| Type | Certified Reference Material |
| Quantity | 1 g |
| Formula Weight | 218.20 |
| Grade | Certified Reference Material |