missing translation for 'onlineSavingsMsg'
Learn More

Triacetin (Glycerol Triacetate), Reagents

Triacetin (Glycerol Triacetate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Labchem Inc CT100200500C

Quantity 500 mL

 View more versions of this product

Catalog No. 01333452


Only null left

Description

Description

Specifications

Chemical Identifiers

102-76-1
218.21
1,2,3-Propanetriol, Glycerol Triacetate
CC(=O)OCC(COC(C)=O)OC(C)=O
C9H14O6
URAYPUMNDPQOKB-UHFFFAOYSA-N
1,3-bis(acetyloxy)propan-2-yl acetate

Specifications

Triacetin (Glycerol Triacetate)
258°C to 259°C
258°C
C9H14O6
1.156
URAYPUMNDPQOKB-UHFFFAOYSA-N
1,3-bis(acetyloxy)propan-2-yl acetate
Reagent
102-76-1
Colorless
Glass Bottle
500 mL
1,2,3-Propanetriol, Glycerol Triacetate
CC(=O)OCC(COC(C)=O)OC(C)=O
218.21
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions