missing translation for 'onlineSavingsMsg'
Learn More

Triacetin (Glycerol Triacetate), Reagents

Catalog No. p-200021282
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333454 120 mL
01333453 1 L
01333452 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01333454 Supplier Labchem Inc Supplier No. CT100200120C
Only null left

Triacetin (Glycerol Triacetate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 102-76-1
Molecular Formula C9H14O6
Molecular Weight (g/mol) 218.21
InChI Key URAYPUMNDPQOKB-UHFFFAOYSA-N
Synonym 1,2,3-Propanetriol, Glycerol Triacetate
IUPAC Name 1,3-bis(acetyloxy)propan-2-yl acetate
SMILES CC(=O)OCC(COC(C)=O)OC(C)=O

Specifications

Boiling Range 258°C to 259°C
Color Colorless
Boiling Point 258°C
Packaging Glass Bottle
Quantity 120 mL
Specific Gravity 1.156
Grade Reagent
Physical Form Liquid
Chemical Name or Material Triacetin (Glycerol Triacetate)