missing translation for 'onlineSavingsMsg'
Learn More

Triacetin, CP, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor T109820LT

Quantity 20 L
Packaging Steel Can

 View more versions of this product

Catalog No. 18614499


Only null left

Description

Description

Specifications

Chemical Identifiers

102-76-1

Specifications

Triacetin
1
Steel Can
Chemically Pure
102-76-1
258°C
20 L
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions